Learn More
ACL Inc 2"x 12" copper strip
Supplier: ACL Inc 8095
Carbon-loaded water-based polyurethane coating for concrete dissipates static discharge for cost-effective static control. Designed for interior concrete floors and unfinished surfaces, this ESD paint is perfectly suited for temporary ESD work spaces, data centers, warehouses, and creating Faraday cages.