Learn More
BioSpec Products 2.0 ml Vial Rack 5/pk
Supplier: BioSpec Products 702VH45
A pack of five polypropylene microvial vial racks. The rack has a standard microplate footprint and has the capacity to hold forty-five 1.5 ml or 2.0 ml microvials on one side or thirty-two 0.5 ml microvials on the other side.