Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Methyl-1,3-dithiane, 97%, Thermo Scientific™
Click to view available options
Quantity:
1mL
5mL
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Density | 1.1200g/mL |
Color | Colorless to Brown |
Assay Percent Range | 97% |
Infrared Spectrum | Authentic |
Molecular Formula | C5H10S2 |
Packaging | Glass bottle |
Quantity | 5mL |
Molecular Weight (g/mol) | 134.255 |
Show More |
RUO – Research Use Only