Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sterilis Solutions 2 Gallon Sharps Container
Sterilis Solutions 2 Gallon Sharps Container - Puncture-resistant sharps container with wide mailbox opening for easy loading of needles.
$238.50 - $870.00
Specifications
Material | Polypropylene |
---|---|
Product Type | Sharps Container |
Certifications/Compliance | ISO23907-1:2019 and ISO23908 |
Closure Type | Mailbox Lid |
Closure Color | White/Translucent |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-777-028
|
Sterilis Solutions
3823343701 |
20/Cs. |
Case of 20 for $238.50
|
|
|||||
23-777-029
|
Sterilis Solutions
3823343702KIT |
40/Cs. |
Case of 40 for $623.00
|
|
|||||
23-777-030
|
Sterilis Solutions
3823343703KIT |
60/Cs. |
Case of 60 for $870.00
|
|
|||||
Description
- Used to comply with FDA and OSHA standards for sharps and biohazard waste collection.
- General-purpose sharps container with clear locking lid and marked with biohazard label.
- Puncture-resistant with wide mailbox slot opening for horizontal loading of needles.
- Made from transparent red puncture-resistant polypropylene that can be autoclaved or incinerated.
- Rotating handle for easy transporting and handling.
- Nests to conserve storage space
Specifications
Polypropylene | |
ISO23907-1:2019 and ISO23908 | |
White/Translucent | |
Red | |
Yes | |
Non-sterile |
Sharps Container | |
Mailbox Lid | |
2 gal. | |
13.3 x 11.4 x 5.8 in. | |
Yes | |
Autoclavable |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title