Learn More
Vector Laboratories 2.5% Normal Goat Serum Blocking Solution 50 ml
Supplier: Vector Laboratories S101250
2.5% Normal Goat Serum is used for blocking non-specific binding when goat secondary antibodies are used in immunohistochemistry assays. A ready-to-use solution, it is the same serum preparation that is included in the ImmPRESS Detection Systems. It can also be used as an antibody diluent. Vector produces this serum by collecting and pooling the sera from healthy adult goats, heat-treating and centrifuging the pool to remove precipitates, and then filtering to remove fine particulates. The serum is tested with antibodies to check for possible cross-reactivities and then diluted to 2.5% (v/v) in buffer containing 0.08% sodium azide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.