Learn More
ProteinSimple 2-40 kDa Separation Module, 8 x 25 capillary cartridges, Pack 5
Supplier: ProteinSimple SMW012
Compatible with Jess, Abby and Wes systems using Compass v3.0 or higher. Includes separation box with Capillary Cartridges, 2-40 kDa Pre-filled Plates (pre-filled with Separation Matrix 2, Stacking Matrix 2, Split Running Buffer 2 and Matrix Removal Buffer), Sample Buffer, Wash Buffer, plus Standard Pack 1 clamshell with DTT, FL Standard master mix with 29 kDa System Control protein.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.