Learn More
Cole-Parmer Reversible 16-Point Vertical Temperature Label, 14-31C/58-88F; 10/Pk
Supplier: Cole-Parmer 0903720
Digi-Sense Reversible 16-Point Vertical Temperature Label, 14-31C/58-88F; 10/Pk, Each style covers a wide range of temperatures, for use in many different applicationNon-toxic / non-hazardous temperature measuring alternativeEasy to read and unbreakable