Learn More
Chemglass Life Sciences #15 Internal Bushing, Solid, PTFE, #210 Viton O-Ring
Supplier: Chemglass Life Sciences CG36303
#15 Internal Bushing, Solid, PTFE, #210 Viton O-Ring. Solid bushing for use with CG-360 internal glass threads. Supplied with one Viton o-ring.