Learn More
COLUMBIA ANALYTICAL INSTRUMENTS INC 1500 PPM EPA COD TUBES 150/PK
Supplier: COLUMBIA ANALYTICAL INSTRUMENTS INC COD1500.150
1500ppm range COD Tubes for compliance testing, high precision means low MDL's. QC Document included.