Learn More
Polysciences, Inc. Polybead Microspheres 15.00 um, 5 mL
Supplier: Polysciences, Inc. 183285
Polybead Microspheres are monodisperse polystyrene microspheres available in sizes from 50nm to 90um. Diameters 4.5um and larger are crosslinked with divinylbenzene (DVB). Our experienced chemists control the synthesis to provide precise monodisperse particle size distributions. These particles contain a slight anionic charge from sulfate ester.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.