Learn More
Cliawaived Inc MultiTox Urine Control (Positive) – 25mL
SDP
Supplier:  Cliawaived Inc MULTITOXPC25E
Encompass
Multi-Tox Urine Control (Positive)- 25mL. FDA Cleared - 16 panel Positive Controls for use with all point of care cups and cards that include THC,COC, PCP, MOR or OPI, AMP, mAMP, BAR, BZO, MTD, MTQ, PPX, TCA, MDMA, OXY, BUP G, and EDDP.
Catalog No. NC1375417
Due to product restrictions, please to purchase or view availability for this product.