Learn More
Beaver Visitec Intl Ltd BLADE 15 DEG 3.0MM DEPTH S/S UBL (6/SP)
Supplier: Beaver Visitec Intl Ltd 377513
Beaver Micro-Sharps and Micro-Unitomes are manufactured using a propriety grinder technology thatyields consistent sharpness. Each blade/knife is color coded for easy identi cation. 3.0 mm, 15 degrees, blue. 6 per box.