Learn More
American International Electric 12IN SEALER
Supplier:  American International Electric AIE300
Encompass_Preferred
IMPULSE HAND SEALERS,Seal Length:12 in. ,Seal Thickness:6 mil,Seal Width:2 mm,Watts:380 W,Weight :11 lbs
Catalog No. NC9593336