Learn More
Chemglass Life Sciences Complete 125mL Bottle, Gas Washing, Tall Form, Coarse Frit, 290mm Approx. OAH
Supplier: Chemglass Life Sciences CG111413
Complete 125mL Bottle, Gas Washing, Tall Form, Coarse Frit, 290mm Approx. OAH. Tall form gas washing bottle having a standard taper 34/28 top joint, hex base and coarse fritted tube for gas dispersion. Stopper is held in place with two CG-110-02, 18mm stainless steel springs. Springs are supplied when bottle is ordered complete.