Learn More
Chemglass Life Sciences 1200mL Style Freeze Dry Flask, P/N 7543000
Supplier: Chemglass Life Sciences CG161108
1200mL Style Freeze Dry Flask, P/N 7543000. Flat bottom borosilicate flask for use with and Virtis freeze dryers. Wide mouth design provides easy access sample.