Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ 11mm Glass Crimp Top Vials - Routine Range
Use these 11mm Glass Crimp Top Vials, Routine Range, for optimum performance in standard applications and high throughput analyses.
Supplier: Thermo Scientific™ 60180ACV111P
Description
Use Thermo Scientific™ 11mm Glass Crimp Top Vials - Routine range, for optimum performance in standard applications and high throughput analyses. These autosampler vials exhibit easy handling and high solvent compatibility for the mainstream HPLC and GC applications including these recommended workflows and applications:
- Chromatographic applications aligning with all pharmacopeias
- Lower risk of contamination, degradation and adsorption
- Standard concentrations
- Perfect autosampler fit
- High throughput analysis
Specifications
2mL vial, clear glass, 11mm crimp top, wide opening, ID patch, 100/Pack | |
Glass | |
Crimp Cap | |
Yes |
Clear | |
2 mL | |
11 mm |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction