Learn More
Chemglass Life Sciences 10mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Medium Frit, GL 14 Thread
Supplier: Chemglass Life Sciences CG186601
10mL Peptide Synthesis Vessel, Solid Phase, T-Bore PTFE Stpk, Vacuum, Medium Frit, GL 14 Thread. Similar to CG-1864, but with the addition of a lower standard taper inner joint to allow collection into a vessel having a 24/40 outer joint. MEDIUM porosity fritted glass resin support. A serrated hose connection is sealed on the inner joint for connection to a vacuum source. The “T”-Bore stopcock has an 8mm O.D. sidearm for connection to nitrogen, while the serrated hose connection has an O.D. of 8mm at the middle ring.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.