Learn More
Chemglass Life Sciences 10mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 255mm OAH
Supplier: Chemglass Life Sciences CG171303
10mL Addition Funnel, Graduated, 24/40 Joint Size, 2mm PTFE Metering Stopcock, 255mm OAH. Cylindrical style addition funnel with a PTFE adjustable bore metering plug. Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow. For replacement stoppers see CG-3000.