Learn More
Foxx Life Sciences Vactrap™ G Vaccum Trap System (Vacuum Flask Alter) for Aspi. and Vacuum Safety, 10+2 L PUREGRIP Glass Bottles Assy /W Secondary Container, 10L Bottle /W 2L Over flow Bottle & Inline Vent Filter, 1/EA
Supplier: Foxx Life Sciences 30RA201FLS
Vactrap™ G (Glass) is the ultimate laboratory vacuum trap system nested neatly in a secondary container with a small footprint and incredible stability against tipping adhering to the strictest CDC recommendations for laboratory safety. The PUREGRIP glass media bottle with extra length EZFlow TPE tubing and tangle-free Polypropylene VersaCap ensures a continuous full vacuum rating. A 0.2μm PTFE inline vent filter retains any aerosolized bacteria and protects the house vacuum or pump from waste produced during supernatant removal, chemical separation, and cell or tissue culture media aspiration. Vactrap™ G combines ease-of-use with the highest temperature rating in the industry, 142°C.•40 CFR 264.175 compliant•3.3 Borosilicate Type 1 Glass•Bleach-Compatible•Autoclavable
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.