Learn More
Weber Scientific 1015-01 CREAM TEST BOTTLES
SDP
Supplier:  Weber Scientific 101501
Encompass
Size 20%. Butyrometer. Subdivision 0.2. Sample Size 5gram. Inaccuracy +/-0.10.
Catalog No. NC0582906