Learn More
Chemglass Life Sciences 100mL Peptide Synthesis Vessel, Solid Phase, Luer, Medium Frit, GL 25 Thread
Supplier: Chemglass Life Sciences CG186204
100mL Peptide Synthesis Vessel, Solid Phase, Luer, Medium Frit, GL 25 Thread. Similar to CG-1860, but with the addition of a Luer inner joint below the 2mm PTFE stopcock. Vessels are available in sizes other than those listed. Please call for details.