Learn More
Chemglass Life Sciences Funnel, 1000mL, Separatory, Blank, Stopper Neck, Size # 27, Glassblowers
Supplier: Chemglass Life Sciences CG62406
Funnel, 1000mL, Separatory, Blank, Stopper Neck, Size # 27, Glassblowers. Squibb shaped separatory blank complete with tooled and finished ground stopper neck top. Lower portion of blank is unfinished and requires a stopcock to be sealed. NOTE: SUPPLIED WITHOUT STOPPER.