Learn More
Chemglass Life Sciences 1000mL Peptide Synthesis Reaction Vessel, Jacketed, Lower Drain, Coarse Frit, 100mm ID Flange, Drain Valve Type/CG-511-01, 100mm Body I.D. x 185mm Height
Supplier: Chemglass Life Sciences CG186804
1000mL Peptide Synthesis Reaction Vessel, Jacketed, Lower Drain, Coarse Frit, 100mm ID Flange, Drain Valve Type/CG-511-01, 100mm Body I.D. x 185mm Height. Cylindrical, jacketed, peptide synthesis reaction vessel having a coarse porosity fritted resin support. The vessel is constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the corresponding lids. Lower outlet is a CG-511 bottom drain valve. Cooling jacket is equipped with two removable hose connections to reduce the risk of injury while attaching or detaching flexible tubing. Supplied complete with two polypropylene hose connections and two GL-14 screw caps. For the clamp see CG-141 and for the o-ring see CG-147. Lid Not Included.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.