Learn More
Microtech Scientific Coliform Bottle, 1000ML, 64/Case
Supplier:  Microtech Scientific MS5RT1000WPEN1
Encompass_Preferred
Water Sample Bottles, Sterile, 1000mL NNI HDPE Bottles, Sterile, NNI Natural Cap, NO Sod. Thio., Paper Tamper Seal, 64/cs, Lot
Catalog No. NC1683802