Learn More
Electron Microscopy Sciences 1% Uranyl Acetate Solution, 100ml
Supplier: Electron Microscopy Sciences 224001
Uranyl acetate is extensively used as a negative stain in electron miroscopy. 1% uranyl acetate solutions is used as anindicator, and atitrantin stronger concentrations inanalytical chemistry, as it forms an insoluble salt withsodium(the vast majority of sodium salts are water-soluble). Uranyl acetate solutions show evidence of being sensitive to light, especially UV, and will precipitate if exposed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.