Learn More
ATLANTIC BIOPHARM SOLUTIONS LLC 1-PIECE 40 SERIES BALL VALVE
Supplier:  ATLANTIC BIOPHARM SOLUTIONS LLC ABSSS45S8
Encompass_Preferred
Stainless Steel 1-Piece 40 Series Ball Valve, 12 Cv, 1/2 in. compression ends
Catalog No. NC1751330