Learn More
Cayman Chemical 1-Methyladenosine, 15763-06-1, 25 mg
Supplier: Cayman Chemical 1693725
A modified nucleoside generated by the processing of tRNA by methyltransferases; urinary and serum levels are elevated in several forms of cancer, supporting its use as a biomarker