Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1-Methyl-2-Pyrrolidinone, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
pH | 8.5 to 10 (at 100.00g/L 20°C) |
Melting Point | -24°C |
Color | Colorless |
Density | 1.028g/mL at 25°C |
Boiling Point | 204°C (1013 hPa) |
Quantity | 6 x 1 L |
Explosion Limit | 0.095 |
Flash Point | 91°C (195.8°F) |
Ignition Point | 518°F |
Odor | Amine-like |
Show More |
Safety and Handling
Hazard Statement | P201-P280-P305 + P351 + P338-P308 + P313 |
RTECSNumber | 212-828-1 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction