Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1% CH4, CO, CO2, C2H2, C2H4 & C2H6 in N2 Analytical Standard, MilliporeSigma™ Supelco™
1% (w/w) each component in nitrogen
Supplier: MilliporeSigma Supelco 23462
Specifications
Gas | |
Analytical Standard | |
48 L |
UN 1066 2.2 | |
Cylinder | |
Not Applicable |
Safety and Handling
H360 - H373
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title