Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ 1-Butanol
SCI_ED

Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Reagent Grade
71-36-3
C4H10O
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO
71-36-3
C4H10O
74.123
LRHPLDYGYMQRHN-UHFFFAOYSA-N
1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
263
CHEBI:28885
butan-1-ol
CCCCO

This product is for educational use - no C of A is available.

Color Colorless
Assay Percent Range >99%
Linear Formula CH3(CH2)2CH2OH
Formula Weight 74.12
Grade Reagent
Density 0.81g/mL
Quantity 4 L
Packaging Reagent Grade
Chemical Name or Material 1-Butanol

ShelfLife : 2 years

DOTInformation : DOT Class 3, III, : Flammable Liquid

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit