Learn More
MSC Accupro 12182388 1/8", 1/2" Length of Cut, 1/8" Shank Diam, 1-1/2" Overall Length, 4 Flute, Solid Carbide Square End Mill Single End, AlTiN Finish, Spiral Flute, 40° Helix, Centercutting, Right Hand Cut, Right Hand Flute
Supplier: MSC 01982222
Accupro 12182388 1/8", 1/2" Length of Cut, 1/8" Shank Diam, 1-1/2" Overall Length, 4 Flute, Solid Carbide Square End Mill Single End, AlTiN Finish, Spiral Flute, 40° Helix, Centercutting, Right Hand Cut, Right Hand Flute