Learn More
American Biotech Supply Premier Countertop Refrigerator Freestanding-LEFT HINGED
Supplier: American Biotech Supply ABTHCUCFS0104GLH
1 Cu. Ft. Capacity. One glass swing door, left hinged. Microprocessor temperature controller. Digital temperature display. Audible and visual high and low temperature alarm.