Learn More
Growcells 0.9% (NaCl)S Sterile Saline
Supplier: Growcells MSDW01004
0.9% Sodium chloride aqueous solution, 18.2 megOhm water, 0.2 micron sterile filtered and steam sterilized in final packaging.