Learn More
Sarstedt Inc Reaction Tube, 0.5 mL, Conical Base, Polypropylene, 1000/Pack
Supplier: Sarstedt Inc 72.699
Roche; Sarstedt; Highly processive 5-3' DNA polymerase lacking 3-5' exonuclease activity; Stable during prolonged incubations at elevated temp (+95deg.C); Highest activity at pH: 9, temp: +75deg.C; 1U/uL; 100U