Learn More
ALLIED BIOMEDICAL TECHNOLOGY INC 0.45% SODIUM CHL INJ CS/12
Supplier:  ALLIED BIOMEDICAL TECHNOLOGY INC 798509
Encompass
EACH 100 mL CONTAINS SODIUMCHLORIDE 450 mg IN WATER FORINJECTION. ELECTROLYTES PER1000 mL: SODIUM 77 mEq;CHLORIDE 77 mEq.154 mOsmol/LITER (CALC.)
Catalog No. NC1441000
Due to product restrictions, please to purchase or view availability for this product.