This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Mutation Detection Assays
›
Search Tool
›
Search Results
› Hs00001559_mu
See other HNF1A Mutant allele assays›
Gene:
HNF1A
Transcript:
NM_000545.6
Assay Name:
HNF1A_21471_mu
COSMIC ID:
21471
Nucleotide Mutation:
c.617G>T
Amino Acid Change
p.W206L
Mutation genome location:
chr.12 120993610 on build GRCh38
Assay gene location:
Within Exon 3
Assay transcription exon location:
Within Exon 3
Assay ID
Hs00001559_mu
Size
75 rxns
75 rxns
Availability
Inventoried
Catalog #
4465804
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
More Information
Genomic Map
LOADING...
×
Back To Top
More Information
Related Assays:
Assay Name
Assay ID
HNF1A_rf
Hs00001371_rf
Back To Top
Related Products
TaqMan
®
Digital PCR Chip Kit
TaqMan
®
Digital PCR Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-blue:8080/10.253.228.45:8080.
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0