This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Mutation Detection Assays
›
Search Tool
›
Search Results
› Hs00001552_mu
See other ERBB4 Mutant allele assays›
Gene:
ERBB4
Transcript:
NM_005235.2
Assay Name:
ERBB4_48363_mu
COSMIC ID:
48363
Nucleotide Mutation:
c.1853A>C
Amino Acid Change
p.H618P
Mutation genome location:
chr.2 211665341 on build GRCh38
Assay gene location:
Within Exon 17
Assay transcription exon location:
Within Exon 15
Assay ID
Hs00001552_mu
Size
75 rxns
75 rxns
Availability
Inventoried
Catalog #
4465804
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
More Information
Genomic Map
LOADING...
×
Back To Top
More Information
Related Assays:
Assay Name
Assay ID
ERBB4_rf
Hs00001367_rf
Back To Top
Related Products
TaqMan
®
Digital PCR Chip Kit
TaqMan
®
Digital PCR Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-1-prod-blue:8080/10.253.225.184:8080.
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0