This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Mutation Detection Assays
›
Search Tool
›
Search Results
› Hs00001365_rf
Gene:
CSF1R
Transcript:
NM_005211.3
Assay Name:
CSF1R_rf
Assay genome location:
chr.5 150060898 on build GRCh38
Assay gene location:
Within Exon 14
Assay transcription exon location:
Within Exon 14
Assay ID
Hs00001365_rf
Size
75 rxns
75 rxns
Availability
Inventoried
Catalog #
4465807
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
LOADING...
×
Back To Top
Related Products
TaqMan
®
Digital PCR Chip Kit
TaqMan
®
Digital PCR Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-blue:8080/10.253.228.45:8080.
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0