This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Mutation Detection Assays
›
Search Tool
›
Search Results
› Hs00001301_mu
See other FLT3 Mutant allele assays›
Gene:
FLT3
Transcript:
NM_004119.2
Assay Name:
FLT3_850_mu
COSMIC ID:
850
Nucleotide Mutation:
c.2520_2521insGGATCC
Amino Acid Change
p.S840_N841insGS
Mutation genome location:
chr.13 28018487-28018488 on build GRCh38
Assay gene location:
Overlaps Intron 23 - Exon 23
Assay ID
Hs00001301_mu
Size
75 rxns
75 rxns
Availability
Inventoried
Catalog #
4465804
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
More Information
Genomic Map
LOADING...
×
Back To Top
More Information
Related Assays:
Assay Name
Assay ID
FLT3_rf
Hs00001016_rf
Back To Top
Related Products
TaqMan
®
Digital PCR Chip Kit
TaqMan
®
Digital PCR Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-blue:8080/10.253.228.45:8080.
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0