This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Mutation Detection Assays
›
Search Tool
›
Search Results
› Hs00000824_mu
See other PIK3CA Mutant allele assays›
Gene:
PIK3CA
Transcript:
NM_006218.3
Assay Name:
PIK3CA_763_mu
COSMIC ID:
763
Nucleotide Mutation:
c.1633G>A
Amino Acid Change
p.E545K
Mutation genome location:
chr.3 179218303 on build GRCh38
Assay gene location:
Overlaps Exon 12 - Intron 12
Assay ID
Hs00000824_mu
Size
75 rxns
75 rxns
Availability
Inventoried
Catalog #
4465804
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
More Information
Genomic Map
LOADING...
×
Back To Top
More Information
Related Assays:
Assay Name
Assay ID
PIK3CA_rf
Hs00001025_rf
Back To Top
Related Products
TaqMan
®
Digital PCR Chip Kit
TaqMan
®
Digital PCR Master Mix
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-srch-3-prod-blue:8080/10.253.228.45:8080.
git-commit: 9c2c959b77162a04e431fec2cb3661656af8b157
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.4.0-2024.02.08-1.0