Assay Details
Amplicon Length: | 109 |
Chromosome Location: | Chr. 15 - 74788928 - 74788928 [-] on 74788928 Build GRCh38 |
Species: | Human |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-4513 | MIMAT0019050 |
Stem-loop Details
Human