Assay Details
Target Gene Details
Entrez Gene ID: | 14673 |
Gene Name: | guanine nucleotide binding protein, alpha 12 |
Gene Aliases: |
AI414047, AI504261, Galpha12 |
Location: |
Chr.5:140759944-140830431 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gna12 | NM_010302.2 | 1 | 275 | NP_034432.1 |
BC111810.1 | AAI11811.1 | |||
M63659.1 | 1 | 275 | AAA37648.1 |
More Information
Set Membership: |
Intragenic Exonic |