Assay Details
Target Gene Details
Entrez Gene ID: | 50780 |
Gene Name: | regulator of G-protein signaling 3 |
Gene Aliases: |
4930506N09Rik, C2PA-RGS3, C2pa, PDZ-RGS3, RGS3S |
Location: |
Chr.4:62559838-62703022 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 25 - Exon 26 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs3 | NM_134257.3 | NP_599018.3 | ||
AF215669.1 | AAF34626.1 | |||
AF215670.1 | AAF34627.1 | |||
AF350047.1 | AAK38878.1 | |||
AK004648.1 | BAB23439.1 | |||
AK084729.1 | BAC39265.1 | |||
AK158257.1 | BAE34430.1 | |||
AK171666.1 | BAE42598.1 | |||
AK199800.1 | ||||
BC033449.1 | AAH33449.1 |
More Information
Set Membership: |
Intragenic Non-exonic |