Assay Details
Target Gene Details
Entrez Gene ID: | 13992 |
Gene Name: | KH domain containing, RNA binding, signal transduction associated 3 |
Gene Aliases: |
Etle, SLM-2, Salp, Slm2, T-STAR |
Location: |
Chr.15:68928405-69101163 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Khdrbs3 | NM_010158.2 | 1 | 96 | NP_034288.2 |
AF079763.1 | 1 | 98 | AAC31753.1 | |
AF099092.1 | 1 | 98 | AAC72396.1 | |
AK014353.1 | 1 | 85 | ||
BC057577.1 | 1 | 80 | AAH57577.1 |
More Information
Set Membership: |
Intragenic Exonic |