Assay Details
Target Gene Details
Entrez Gene ID: | 16341 |
Gene Name: | eukaryotic translation initiation factor 3, subunit E |
Gene Aliases: |
48kDa, Eif3s6, Int6, eIF3-p46, eIF3-p48 |
Location: |
Chr.15:43250040-43282736 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Eif3e | NM_008388.2 | NP_032414.1 | ||
AK002576.1 | ||||
AK011436.1 | BAB27621.1 | |||
AK088104.1 | ||||
AK088269.1 | ||||
AK145059.1 | BAE26211.1 | |||
AK146936.1 | BAE27546.1 | |||
AK161650.1 | BAE36512.1 | |||
AK165929.1 | BAE38465.1 | |||
BC029177.1 | AAH29177.1 |
More Information
Set Membership: |
Intragenic Non-exonic |