Assay Details
Target Gene Details
Entrez Gene ID: | 16798 |
Gene Name: | large tumor suppressor |
Gene Aliases: |
AW208599 |
Location: |
Chr.10:7681209-7716461 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Lats1 | NM_010690.1 | NP_034820.1 | ||
AK038612.1 | BAC30063.1 | |||
BC158092.1 | ||||
BC158123.1 |
More Information
Set Membership: |
Intragenic Non-exonic |