Assay Details
Target Gene Details
Entrez Gene ID: | 20758 |
Gene Name: | small proline-rich protein 2D |
Gene Aliases: |
- |
Location: |
Chr.3:92339140-92340873 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sprr2d | NM_011470.2 | NP_035600.1 | ||
AJ005562.1 | CAA06591.1 | |||
AK008778.1 | BAC25232.1 | |||
AY158988.1 | AAN86825.1 |