Assay Details
Target Gene Details
Entrez Gene ID: | 382985 |
Gene Name: | ribonucleotide reductase M2 B (TP53 inducible) |
Gene Aliases: |
p53R2 |
Location: |
Chr.15:37923952-37961108 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rrm2b | NM_199476.1 | 9 | 3104 | NP_955770.1 |
BC058103.1 | 9 | 3104 | AAH58103.1 |
More Information
Set Membership: |
Intragenic Exonic |