Assay Details
Target Gene Details
Entrez Gene ID: | 20308 |
Gene Name: | chemokine (C-C motif) ligand 9 |
Gene Aliases: |
CCF18, MRP-2, Scya10, Scya9 |
Location: |
Chr.11:83572917-83578636 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccl9 | NM_011338.2 | 4 | 709 | NP_035468.1 |
AK151131.1 | 4 | 577 | BAE30139.1 | |
AK151340.1 | 1 | 266 | BAE30318.1 | |
AK151649.1 | 4 | 577 | BAE30578.1 | |
AK154511.1 | 4 | 577 | BAE32640.1 | |
AK154657.1 | 4 | 577 | BAE32747.1 | |
AK155032.1 | 4 | 577 | BAE33004.1 | |
AK155036.1 | 4 | 577 | BAE33005.1 | |
BC139318.1 | ||||
BC145962.1 | ||||
BE626870.1 | ||||
U15209.1 | 4 | 690 | AAA92583.1 | |
U19482.1 | 4 | 589 | AAB17120.1 | |
U49513.1 | 4 | 565 | AAB02198.1 |
More Information
Set Membership: |
Intragenic Exonic |