Assay Details
Target Gene Details
Entrez Gene ID: | 73660 |
Gene Name: | calcium binding protein 4 |
Gene Aliases: |
2410038D05Rik |
Location: |
Chr.19:4135423-4144080 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cabp4 | NM_144532.2 | 1 | 336 | NP_653115.1 |
AK171863.1 | 5 | 946 | ||
AY039218.1 | 1 | 331 | AAK83463.1 | |
BC049263.1 | 1 | 336 | AAH49263.1 |